accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNH2N

CAS RN 21109-25-1

IUPAC Name: 1-(1H-indol-2-yl)methanamine
Synonyms: 1-(1H-indol-2-yl)methanamine
Molecular Weight (g/mol): 146.19
Molecular Formula: C9H10N2
InChi Key: RNAODKZCUVVPEN-UHFFFAOYSA-N
SMILES: NCC1=CC2=CC=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results