accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21124-13-0

H3CNNNHCH3(E/Z)

CAS RN 21124-13-0

IUPAC Name: 1-methyl-3-(4-methylphenyl)triaz-1-ene
Synonyms: 1-methyl-3-(4-methylphenyl)triaz-1-ene
Molecular Weight (g/mol): 149.2
Molecular Formula: C8H11N3
InChi Key: DNGJVDGPCGXBFF-UHFFFAOYSA-N
SMILES: CN=NNC1=CC=C(C)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results