CAS RN 2113-56-6
IUPAC Name:
3-methoxy-1,1'-biphenyl
Synonyms:
3-methoxy-1,1'-biphenyl
Molecular Weight (g/mol):
184.24
Molecular Formula:
C13H12O
InChi Key:
KQMIWCAOEFUBQK-UHFFFAOYSA-N
SMILES:
COC1=CC=CC(=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
3-Methoxybiphenyl 96.0+%, TCI America™
CAS: 2113-56-6 Molecular Formula: C13H12O Molecular Weight (g/mol): 184.24 MDL Number: MFCD00027514 InChI Key: KQMIWCAOEFUBQK-UHFFFAOYSA-N PubChem CID: 257971 IUPAC Name: 3-methoxy-1,1'-biphenyl SMILES: COC1=CC=CC(=C1)C1=CC=CC=C1