accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21154-48-3

IUPAC Name: tetrakis(propan-2-yloxy)germane
Synonyms: tetraisopropoxygermane
Molecular Weight (g/mol): 308.98
Molecular Formula: C12H28GeO4
InChi Key: PKLMYPSYVKAPOX-UHFFFAOYSA-N
SMILES: CC(C)O[Ge](OC(C)C)(OC(C)C)OC(C)C
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)