accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOOOONHNHN(S)

CAS RN 2116-83-8

IUPAC Name: 2,5-dioxopyrrolidin-1-yl (2S)-2,6-di{[(benzyloxy)carbonyl]amino}hexanoate
Synonyms: 2,5-dioxopyrrolidin-1-yl (2S)-2,6-di{[(benzyloxy)carbonyl]amino}hexanoate
Molecular Weight (g/mol): 511.53
Molecular Formula: C26H29N3O8
InChi Key: LHOAUCZIIQFZMI-NRFANRHFSA-N
SMILES: O=C(NCCCC[C@H](NC(=O)OCC1=CC=CC=C1)C(=O)ON1C(=O)CCC1=O)OCC1=CC=CC=C1
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)