accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3CCH3OSiOSiCH3H3CH3COSiH3CCH3CH3

CAS RN 2116-84-9

IUPAC Name: 2,2,6,6-tetramethyl-4-phenyl-4-[(trimethylsilyl)oxy]-3,5-dioxa-2,4,6-trisilaheptane
Synonyms: phenyl trimethicone
Molecular Weight (g/mol): 372.76
Molecular Formula: C15H32O3Si4
InChi Key: LINXHFKHZLOLEI-UHFFFAOYSA-N
SMILES: C[Si](C)(C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results