accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCH3O

CAS RN 21170-10-5

IUPAC Name: 3a,6a-dimethyl-octahydropentalene-2,5-dione
Synonyms: 3a,6a-dimethyl-tetrahydropentalene-2,5-dione
Molecular Weight (g/mol): 166.22
Molecular Formula: C10H14O2
InChi Key: KGZWVKASYOPUET-UHFFFAOYSA-N
SMILES: CC12CC(=O)CC1(C)CC(=O)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results