CAS RN 211737-28-9
IUPAC Name:
2-bromo-5-hexylthiophene
Synonyms:
2-bromo-5-hexylthiophene
Molecular Weight (g/mol):
247.19
Molecular Formula:
C10H15BrS
InChi Key:
LSIYDKGTGHFTFW-UHFFFAOYSA-N
SMILES:
CCCCCCC1=CC=C(Br)S1
1
–
1
of
1
results
1
2-Bromo-5-hexylthiophene 98.0+%, TCI America™
CAS: 211737-28-9 Molecular Formula: C10H15BrS Molecular Weight (g/mol): 247.194 MDL Number: MFCD10000906 InChI Key: LSIYDKGTGHFTFW-UHFFFAOYSA-N PubChem CID: 10562418 IUPAC Name: 2-bromo-5-hexylthiophene SMILES: CCCCCCC1=CC=C(S1)Br