CAS RN 21190-93-2
CAS RN 21190-93-2
IUPAC Name:
1-(6-methoxypyridin-2-yl)ethan-1-one
Synonyms:
1-(6-methoxypyridin-2-yl)ethanone
Molecular Weight (g/mol):
151.17
Molecular Formula:
C8H9NO2
InChi Key:
XPJALZRCJQKJKB-UHFFFAOYSA-N
SMILES:
COC1=CC=CC(=N1)C(C)=O
1
–
1
of
1
results
1
2-Acetyl-6-methoxypyridine 97.0+%, TCI America™
CAS: 21190-93-2 Molecular Formula: C8H9NO2 Molecular Weight (g/mol): 151.17 MDL Number: MFCD08276836 InChI Key: XPJALZRCJQKJKB-UHFFFAOYSA-N PubChem CID: 13642086 IUPAC Name: 1-(6-methoxypyridin-2-yl)ethan-1-one SMILES: COC1=CC=CC(=N1)C(C)=O