accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 212079-30-6

IUPAC Name: N-{8-methyl-8-azabicyclo[3.2.1]octan-3-ylidene}hydroxylamine hydrochloride
Synonyms: N-{8-methyl-8-azabicyclo[3.2.1]octan-3-ylidene}hydroxylamine hydrochloride
Molecular Weight (g/mol): 190.67
Molecular Formula: C8H15ClN2O
InChi Key: XEGNNKBCZMXBOA-UHFFFAOYNA-N
SMILES: Cl.CN1C2CCC1CC(C2)=NO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results