accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21210-90-2

IUPAC Name: 2-[5-(thiophen-2-yl)thieno[3,2-b]thiophen-2-yl]thiophene
Synonyms: 2-[5-(thiophen-2-yl)thieno[3,2-b]thiophen-2-yl]thiophene
Molecular Weight (g/mol): 304.46
Molecular Formula: C14H8S4
InChi Key: FDQXVHKNZBLBFY-UHFFFAOYSA-N
SMILES: S1C=CC=C1C1=CC2=C(S1)C=C(S2)C1=CC=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results