accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21211-22-3

IUPAC Name: 3-chloro-1-benzothiophene-2-carboxylic acid
Synonyms: 3-chloro-1-benzothiophene-2-carboxylic acid
Molecular Weight (g/mol): 212.65
Molecular Formula: C9H5ClO2S
InChi Key: HJTMIYKPPPYDRJ-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Cl)C2=CC=CC=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results