accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21211-65-4

IUPAC Name: 2-[(1H-pyrrol-2-yl)methyl]-1H-pyrrole
Synonyms: dipyrromethane 2,2'-dipyrrolylmethane pyrromethane dipyrrylmethane 5,10-dihydrodipyrrin
Molecular Weight (g/mol): 146.19
Molecular Formula: C9H10N2
InChi Key: PBTPREHATAFBEN-UHFFFAOYSA-N
SMILES: C(C1=CC=CN1)C1=CC=CN1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results