accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21230-07-9

IUPAC Name: 4-methyl-N-(propan-2-yl)benzene-1-sulfonamide
Synonyms: N-isopropyl-4-methylbenzenesulfonamide
Molecular Weight (g/mol): 213.3
Molecular Formula: C10H15NO2S
InChi Key: NXUHAAIJVABUMJ-UHFFFAOYSA-N
SMILES: CC(C)NS(=O)(=O)C1=CC=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results