accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 212322-56-0

IUPAC Name: ethyl 3-{1-[3-amino-4-(methylamino)phenyl]-N-(pyridin-2-yl)formamido}propanoate
Synonyms: ethyl 3-{1-[3-amino-4-(methylamino)phenyl]-N-(pyridin-2-yl)formamido}propanoate
Molecular Weight (g/mol): 342.4
Molecular Formula: C18H22N4O3
InChi Key: PCPATNZTKBOKOY-UHFFFAOYSA-N
SMILES: CCOC(=O)CCN(C(=O)C1=CC(N)=C(NC)C=C1)C1=CC=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results