CAS RN 212385-73-4
IUPAC Name:
9-{4'-bromo-[1,1'-biphenyl]-4-yl}-9H-carbazole
Synonyms:
9-{4'-bromo-[1,1'-biphenyl]-4-yl}carbazole
Molecular Weight (g/mol):
398.3
Molecular Formula:
C24H16BrN
InChi Key:
OGENPBMBOLTWLZ-UHFFFAOYSA-N
SMILES:
BrC1=CC=C(C=C1)C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
1
–
1
of
1
results
1
9-(4'-Bromo-4-biphenylyl)carbazole 98.0+%, TCI America™
CAS: 212385-73-4 Molecular Formula: C24H16BrN Molecular Weight (g/mol): 398.30 MDL Number: MFCD27644692 InChI Key: OGENPBMBOLTWLZ-UHFFFAOYSA-N PubChem CID: 58892386 IUPAC Name: 9-{4'-bromo-[1,1'-biphenyl]-4-yl}-9H-carbazole SMILES: BrC1=CC=C(C=C1)C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2