accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOHONH

CAS RN 212688-52-3

IUPAC Name: 9-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)nonanoic acid
Synonyms: 9-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}nonanoic acid
Molecular Weight (g/mol): 395.5
Molecular Formula: C24H29NO4
InChi Key: LWISAKTUHDLUTN-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCCCNC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)