accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21279-62-9

IUPAC Name: 3-chloropyrazine-2-carboxamide
Synonyms: 3-chloropyrazine-2-carboxamide
Molecular Weight (g/mol): 157.56
Molecular Formula: C5H4ClN3O
InChi Key: YHPMRHPLAQSPHJ-UHFFFAOYSA-N
SMILES: NC(=O)C1=NC=CN=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results