accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHBrBrOH(E)

CAS RN 21285-46-1

IUPAC Name: (2E)-2,3-dibromobut-2-ene-1,4-diol
Synonyms: (2E)-2,3-dibromobut-2-ene-1,4-diol
Molecular Weight (g/mol): 245.9
Molecular Formula: C4H6Br2O2
InChi Key: MELXIJRBKWTTJH-ONEGZZNKSA-N
SMILES: OC\C(Br)=C(/Br)CO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)