accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21286-54-4

IUPAC Name: [(1S,4R)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonyl chloride
Synonyms: [(1S,4R)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonyl chloride
Molecular Weight (g/mol): 250.74
Molecular Formula: C10H15ClO3S
InChi Key: BGABKEVTHIJBIW-GMSGAONNSA-N
SMILES: CC1(C)[C@@H]2CC[C@@]1(CS(Cl)(=O)=O)C(=O)C2
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results