accessibility menu, dialog, popup

UserName

Viewing profile as user:
NS

CAS RN 21298-53-3

IUPAC Name: 3-(thiophen-2-yl)pyridine
Synonyms: 3-(thiophen-2-yl)pyridine
Molecular Weight (g/mol): 161.22
Molecular Formula: C9H7NS
InChi Key: ZHBLIWDUZHFSJW-UHFFFAOYSA-N
SMILES: S1C=CC=C1C1=CN=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results