accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21298-55-5

SN

CAS RN 21298-55-5

IUPAC Name: 2-(thiophen-3-yl)pyridine
Synonyms: 2-(thiophen-3-yl)pyridine
Molecular Weight (g/mol): 161.22
Molecular Formula: C9H7NS
InChi Key: MINKMXOCFYGGKA-UHFFFAOYSA-N
SMILES: S1C=CC(=C1)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results