accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 213211-69-9

IUPAC Name: (2-ethoxyphenyl)boronic acid
Synonyms: 2-ethoxyphenylboronic acid
Molecular Weight (g/mol): 165.98
Molecular Formula: C8H11BO3
InChi Key: DGFCTCGCMKEILT-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC=C1B(O)O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results