accessibility menu, dialog, popup

UserName

Viewing profile as user:
PFFFFFFNa

CAS RN 21324-39-0

IUPAC Name: sodium hexafluoro-λ⁵-phosphanuide
Synonyms: sodium hexafluorophosphate
Molecular Weight (g/mol): 167.95
Molecular Formula: F6NaP
InChi Key: KMADQUOCJBLXRP-UHFFFAOYSA-N
SMILES: [Na+].F[P-](F)(F)(F)(F)F
Percent Purity 
  • (1)
  • (3)
  • (2)
  • (7)
Quantity 
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (4)
  • (3)
  • (2)
Grade 
  • (3)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (1)
  • (8)
  • (1)
  • (3)
  • (2)
  • (7)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (4)
  • (3)
  • (2)
  • (3)
  • (5)