accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3COONBHOHO

CAS RN 213318-44-6

IUPAC Name: {1-[(tert-butoxy)carbonyl]-1H-indol-2-yl}boronic acid
Synonyms: 1-(tert-butoxycarbonyl)indol-2-ylboronic acid
Molecular Weight (g/mol): 261.08
Molecular Formula: C13H16BNO4
InChi Key: SVIBPSNFXYUOFT-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1C(=CC2=CC=CC=C12)B(O)O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)