accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 213343-65-8

IUPAC Name: λ¹-rhodium(1+) (1Z,5Z)-cycloocta-1,5-diene (2S,5S)-1-{2-[(2S,5S)-2,5-dimethylphospholan-1-yl]ethyl}-2,5-dimethylphospholane tetrafluoroboranuide
Synonyms: λ¹-rhodium(1+) (S,S)-me-bpe 1,5-cyclooctadiene, (Z,Z)- tetrafluoroborate
Molecular Weight (g/mol): 556.22
Molecular Formula: C22H40BF4P2Rh
InChi Key: CVUPEPGZCZHSOF-BDNLEJQTNA-N
SMILES: [Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.C[C@H]1CC[C@H](C)P1CCP1[C@@H](C)CC[C@@H]1C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)