accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)(S)(S)(S)FO(S)H3C(R)F(S)OH(S)H3C(R)HOOHO

CAS RN 2135-17-3

IUPAC Name: (1R,2R,3aS,3bS,5S,9aS,9bR,10S,11aS)-5,9b-difluoro-1,10-dihydroxy-1-(2-hydroxyacetyl)-2,9a,11a-trimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Synonyms: flumethasone flumethasonum 6α-fluorodexamethasone flumethason aniprime fluorodexamethasone cortexilar 氟米松 anaprime flucorticin
Molecular Weight (g/mol): 410.46
Molecular Formula: C22H28F2O5
InChi Key: WXURHACBFYSXBI-GQKYHHCASA-N
SMILES: C[C@@H]1C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)