accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21354-98-3

IUPAC Name: 5-bromo-4-methyl-2-phenyl-1,3-oxazole
Synonyms: 5-bromo-4-methyl-2-phenyl-1,3-oxazole
Molecular Weight (g/mol): 238.08
Molecular Formula: C10H8BrNO
InChi Key: QHQQHNFHCQSTBJ-UHFFFAOYSA-N
SMILES: CC1=C(Br)OC(=N1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results