accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CP

CAS RN 213697-53-1

IUPAC Name: 2'-(dicyclohexylphosphanyl)-N,N-dimethyl-[1,1'-biphenyl]-2-amine
Synonyms: 2'-(dicyclohexylphosphanyl)-N,N-dimethyl-[1,1'-biphenyl]-2-amine
Molecular Weight (g/mol): 393.56
Molecular Formula: C26H36NP
InChi Key: ZEMZPXWZVTUONV-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=CC=C1C1=CC=CC=C1P(C1CCCCC1)C1CCCCC1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)