CAS RN 213743-31-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Lck Inhibitor
The Lck Inhibitor, also referenced under CAS 213743-31-8, controls the biological activity of Lck. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.