accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHClHO

CAS RN 2138-22-9

IUPAC Name: 4-chlorobenzene-1,2-diol
Synonyms: 4-chlorocatechol
Molecular Weight (g/mol): 144.55
Molecular Formula: C6H5ClO2
InChi Key: WWOBYPKUYODHDG-UHFFFAOYSA-N
SMILES: OC1=CC=C(Cl)C=C1O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results