CAS RN 213978-61-1
IUPAC Name:
4-[(4-methoxyphenyl)methoxy]butane-1,2-diol
Synonyms:
4-[(4-methoxyphenyl)methoxy]butane-1,2-diol
Molecular Weight (g/mol):
226.27
Molecular Formula:
C12H18O4
InChi Key:
YRHIJTKHDXQBLC-UHFFFAOYNA-N
SMILES:
COC1=CC=C(COCCC(O)CO)C=C1
1
–
1
of
1
results
1
(R)-4-(4-Methoxybenzyloxy)-1,2-butanediol 96.0+%, TCI America™
CAS: 213978-61-1 Molecular Formula: C12H18O4 Molecular Weight (g/mol): 226.272 InChI Key: YRHIJTKHDXQBLC-LLVKDONJSA-N PubChem CID: 44629947 IUPAC Name: (2R)-4-[(4-methoxyphenyl)methoxy]butane-1,2-diol SMILES: COC1=CC=C(C=C1)COCCC(CO)O