accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3O(R)(R)OH(R)OHO(R)NNOH2N

CAS RN 2140-72-9

IUPAC Name: 4-amino-1-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxyoxolan-2-yl]-1,2-dihydropyrimidin-2-one
Synonyms: 2'-O-methylcytidine O(2')-methylcytidine
Molecular Weight (g/mol): 257.25
Molecular Formula: C10H15N3O5
InChi Key: RFCQJGFZUQFYRF-ZOQUXTDFSA-N
SMILES: CO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=CC(N)=NC1=O
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)