accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3O(R)(R)OH(R)OHO(R)NNNH2NN

CAS RN 2140-79-6

IUPAC Name: (2R,3R,4R,5R)-5-(6-amino-9H-purin-9-yl)-2-(hydroxymethyl)-4-methoxyoxolan-3-ol
Synonyms: (2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-2-(hydroxymethyl)-4-methoxyoxolan-3-ol
Molecular Weight (g/mol): 281.27
Molecular Formula: C11H15N5O4
InChi Key: FPUGCISOLXNPPC-IOSLPCCCSA-N
SMILES: CO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C(N)N=CN=C12
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)