CAS RN 21424-24-8
IUPAC Name:
heptadecyltrimethylazanium bromide
Synonyms:
heptadecyltrimethylazanium bromide
Molecular Weight (g/mol):
378.48
Molecular Formula:
C20H44BrN
InChi Key:
TTXDNWCDEIIMDP-UHFFFAOYSA-M
SMILES:
[Br-].CCCCCCCCCCCCCCCCC[N+](C)(C)C
1
–
1
of
1
results
1
Heptadecyltrimethylammonium Bromide 98.0+%, TCI America™
CAS: 21424-24-8 Molecular Formula: C20H44BrN Molecular Weight (g/mol): 378.483 InChI Key: TTXDNWCDEIIMDP-UHFFFAOYSA-M Synonym: Trimethylheptadecylammonium Bromide PubChem CID: 10045219 IUPAC Name: heptadecyl(trimethyl)azanium;bromide SMILES: CCCCCCCCCCCCCCCCC[N+](C)(C)C.[Br-]