accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOFCl

CAS RN 214262-85-8

IUPAC Name: ethyl 2-(2-chloro-6-fluorophenyl)acetate
Synonyms: ethyl 2-(2-chloro-6-fluorophenyl)acetate
Molecular Weight (g/mol): 216.64
Molecular Formula: C10H10ClFO2
InChi Key: CEWCLHPMDWPDNQ-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=C(F)C=CC=C1Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results