accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOHNOCl

CAS RN 21427-61-2

IUPAC Name: 5-chloro-3-nitro-1,2-dihydropyridin-2-one
Synonyms: 5-chloro-3-nitro-1H-pyridin-2-one
Molecular Weight (g/mol): 174.54
Molecular Formula: C5H3ClN2O3
InChi Key: QVGQNICXNZMXQA-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Cl)=CNC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results