accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COBr

CAS RN 21436-52-2

IUPAC Name: 1-bromohexadecan-2-one
Synonyms: 1-bromohexadecan-2-one
Molecular Weight (g/mol): 319.33
Molecular Formula: C16H31BrO
InChi Key: ICZYLTOWIBJLIK-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCC(=O)CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results