accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2144-37-8

IUPAC Name: methyl 5-(chloromethyl)furan-2-carboxylate
Synonyms: methyl 5-(chloromethyl)furan-2-carboxylate
Molecular Weight (g/mol): 174.58
Molecular Formula: C7H7ClO3
InChi Key: PWXMEBZOKUPCST-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(CCl)O1
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)