accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH2OOFFFFFFFFFFFFF

CAS RN 2144-53-8

IUPAC Name: 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl 2-methylprop-2-enoate
Synonyms: 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl 2-methylprop-2-enoate
Molecular Weight (g/mol): 432.18
Molecular Formula: C12H9F13O2
InChi Key: CDXFIRXEAJABAZ-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)