accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OOHNNN

CAS RN 214472-06-7

IUPAC Name: tert-butyl N-[(2-cyanopyridin-4-yl)methyl]carbamate
Synonyms: tert-butyl N-[(2-cyanopyridin-4-yl)methyl]carbamate
Molecular Weight (g/mol): 233.27
Molecular Formula: C12H15N3O2
InChi Key: RVMVGHFPBFHBTD-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC1=CC(=NC=C1)C#N
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)