accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 214493-03-5

IUPAC Name: 5,5'-dibromo-4,4'-dihexyl-2,2'-bithiophene
Synonyms: 5,5'-dibromo-4,4'-dihexyl-2,2'-bithiophene
Molecular Weight (g/mol): 492.37
Molecular Formula: C20H28Br2S2
InChi Key: MWUMRWQISLASDX-UHFFFAOYSA-N
SMILES: CCCCCCC1=C(Br)SC(=C1)C1=CC(CCCCCC)=C(Br)S1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results