accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3MgBr

CAS RN 21450-64-6

IUPAC Name: bromo(2,6-dimethylphenyl)magnesium
Synonyms: bromo(2,6-dimethylphenyl)magnesium
Molecular Weight (g/mol): 209.37
Molecular Formula: C8H9BrMg
InChi Key: BIGUVDUZGAJWQK-UHFFFAOYSA-M
SMILES: CC1=CC=CC(C)=C1[Mg]Br
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)