accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO(S)OO

CAS RN 21461-84-7

IUPAC Name: (2S)-5-oxooxolane-2-carboxylate
Synonyms: (2S)-5-oxooxolane-2-carboxylate
Molecular Weight (g/mol): 129.09
Molecular Formula: C5H5O4
InChi Key: QVADRSWDTZDDGR-VKHMYHEASA-M
SMILES: [O-]C(=O)[C@@H]1CCC(=O)O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results