accessibility menu, dialog, popup

UserName

Viewing profile as user:
MgBr

CAS RN 21473-01-8

IUPAC Name: bromo(naphthalen-2-yl)magnesium
Synonyms: bromo(naphthalen-2-yl)magnesium
Molecular Weight (g/mol): 231.38
Molecular Formula: C10H7BrMg
InChi Key: XNUCNPPAQXKAOY-UHFFFAOYSA-M
SMILES: Br[Mg]C1=CC=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)