accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21473-02-9

IUPAC Name: bromo(4-phenoxyphenyl)magnesium
Synonyms: bromo(4-phenoxyphenyl)magnesium
Molecular Weight (g/mol): 273.41
Molecular Formula: C12H9BrMgO
InChi Key: DTIKYZPTJRZFTL-UHFFFAOYSA-M
SMILES: Br[Mg]C1=CC=C(OC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results