accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2150-47-2

IUPAC Name: methyl 2,4-dihydroxybenzoate
Synonyms: methyl 2,4-dihydroxybenzoate
Molecular Weight (g/mol): 168.15
Molecular Formula: C8H8O4
InChi Key: IIFCLXHRIYTHPV-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(O)C=C1O
Molecular Weight (g/mol) 
  • (4)
  • (5)
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (3)
  • (4)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (4)
  • (5)
  • (2)
  • (7)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)