accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 215162-92-8

IUPAC Name: 5-isocyanato-2,3-dihydro-1-benzofuran
Synonyms: 5-isocyanato-2,3-dihydro-1-benzofuran
Molecular Weight (g/mol): 161.16
Molecular Formula: C9H7NO2
InChi Key: WAIKTAFMGLVRJC-UHFFFAOYSA-N
SMILES: O=C=NC1=CC2=C(OCC2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results