accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NSSSOOCl

CAS RN 215434-25-6

IUPAC Name: 5-(2-methyl-1,3-thiazol-4-yl)thiophene-2-sulfonyl chloride
Synonyms: 5-(2-methyl-1,3-thiazol-4-yl)thiophene-2-sulfonyl chloride
Molecular Weight (g/mol): 279.77
Molecular Formula: C8H6ClNO2S3
InChi Key: YIKNRCVJBOLZFL-UHFFFAOYSA-N
SMILES: CC1=NC(=CS1)C1=CC=C(S1)S(Cl)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results