accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOO

CAS RN 215460-40-5

IUPAC Name: 4-(oxan-4-yloxy)benzaldehyde
Synonyms: 4-(oxan-4-yloxy)benzaldehyde
Molecular Weight (g/mol): 206.24
Molecular Formula: C12H14O3
InChi Key: SIHCMIVOQNBOFV-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(OC2CCOCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results